Dichloromethane ≥99.8% stabilised for HPLC, Supelco®
Supplier: MilliporeSigma
|
Warning
|
DX0838-6
DX0838P-1
DX0838-1
CADX0838-6EA
214
CAD
CADX0838-6
CADX0838P-1
CADX0838-1
Dichloromethane ≥99.8% stabilised for HPLC, Supelco®
Dichloromethane
HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Formula:
CH₂Cl₂ Boiling Pt: 39.8 °C (1013 hPa) Melting Pt: –95 °C Density: 1.322 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III Merck Index: 12,06140 |
|
Specification Test Results
Assay (GC) | 99.8% min |
Appearance | Clear liquid, free from particulates |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Free halogens | To pass test |
Infrared spectrum | Conforms to standard |
Residue after evaporation | 5ppm max |
Titrable acid | 0.0003meq/g max |
UV Abs. at 235 nm | 1.00AU max |
UV Abs. at 240 nm | 0.35AU max |
UV Abs. at 250 nm | 0.10AU max |
UV Abs. at 260 nm | 0.04AU max |
UV Abs. at 270 nm | 0.01AU max |
UV Abs. at 300 nm | 0.01AU max |
UV Abs. at 350 nm | 0.01AU max |
UV Cut-off | 235nm max |
Water (H2O) | 0.02% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...