Dichloromethane ≥99.9% stabilized, OmniSolv®, Supelco®

Supplier: MilliporeSigma

Synonyms: Methylene chloride

DX0830-1 DX0830-1
CADX0830-1CS 1924.75 CAD
CADX0830-1 EM-DX0830-1
Dichloromethane ≥99.9% stabilized, OmniSolv®, Supelco®
Dichloromethane

Stabilized with alkenes. For molecular biology (biosynthesis). UV cutoff 231nm. Lot analysis on label.

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
MDL Number: MFCD00000881
CAS Number: 75-09-2
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://ca.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg

Order Now

Specification Test Results

Max. UV Absorbance at 231 nm 1
Max. UV Absorbance at 240 nm 0.15
Max. UV Absorbance at 250 nm 0.01
Max. UV Absorbance at 270 nm 0.005
Assay (by GC) 99.9% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR