n-Decane ≥98%, Supelco®
Supplier: MilliporeSigma
DX0030-3
DX0030-30
DX0030-30
CADX0030-30EA
10855.1
CAD
CADX0030-30
EM-DX0030-30
CADX0030-3
n-Decane ≥98%, Supelco®
n-Decane
Clear liquid. The infrared spectrum conforms to the standard. Partially soluble in water. Flammable and toxic effects.
Formula:
H₃C(CH₂)₈CH₃ MW: 142.28 g/mol Boiling Pt: 174.1 °C (1013 hPa) Melting Pt: –30 °C Density: 0.73 g/cm³ (20 °C) Flash Pt: 46 °C |
MDL Number:
MFCD00008954 CAS Number: 124-18-5 UN: 2247 ADR: 3,III |
Specification Test Results
Assay (GC) | 98% min |
Color (APHA) | 10 max |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Melting Point | –30°C (–22°F) |
Vapor Density | 4.9 kg/L |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...