Iodine 0.02M in THF/Pyridine/Water (89.6/0.4/10) Oxidizing reagent, Biosynthesis

Supplier: MilliporeSigma

BI0422-0450 BI0422-0950 BI0422-4000
CABI0422-0450CS 1022 CAD
CABI0422-0450 CABI0422-0950 CABI0422-4000
Iodine 0.02M in THF/Pyridine/Water (89.6/0.4/10) Oxidizing reagent, Biosynthesis
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
MDL Number: MFCD00011355
CAS Number: 7553-56-2
Merck Index: 13,05036

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR