Dichloroacetic acid (DCA) 10% in toluene deblocking reagent
Supplier: MilliporeSigma
Synonyms:
DCA
, DCA Deblock
BI0841-NP20
BI0841-NP20
BI0841-4000
BI0841-4000
CABI0841-NP20EA
4054.6
CAD
CABI0841-NP20
CA-BI0841-4000
EM-BI0841-NP20
EM-BI0841-4000
Dichloroacetic acid (DCA) 10% in toluene deblocking reagent
Dichloroacetic acid (DCA)
These deblocking reagents are mainly involved in DNA synthesis.
- Infrared spectrum conforms to the standard
- Corrosive and toxic
- Clear liquid form
Formula:
C₂H₂Cl₂O₂ MW: 128.94 g/mol |
MDL Number:
MFCD00004223 CAS Number: 79-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...