D-(+)-Glucose monohydrate, Millipore®

Supplier: MilliporeSigma

4074-2 4074-4
CA4074-2EA 233 CAD
CA4074-2 CA4074-4
D-(+)-Glucose monohydrate, Millipore®
D(+)-Glucose monohydrate

For Biochemistry.

Formula: C₆H₁₂O₆·H₂O
MDL Number: MFCD00149450
CAS Number: 14431-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Heavy metals (as Pb) 0.001% max
Identity (IR-spectrum) Conforms
Maltose (TLC) 0.2% max
Specific rotation [a]20/D (c=10, water, anhydrous) +52.5-+53.0°
Water (H2O) 10% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR