N-DEX™ Ambidextrous Powder-Free Nitrile Gloves, Showa
Supplier: Showa
N-DEX™
7705PFTM 9905PFM B6005PFL B6005PFM 7705PFTXL B6005PFS 8005PFM 9905PF-XL 9905PFXL 9500PFXL-10 8005PFS 7705PFTS 9905PF-M 9500PFS 8005PFXL 8005PFL B9905PFL 7705PFT-L 6005PF-L 6005PFL 9905PFXS 7705PFT-XL 9905PF-L B9905PFM 6005PFXL B9905PFXS 7705PFT-XS 6005PFM 9500PFL 8005PF-M 9500PFS-07 9500PFL-09 7705PFTL 6005PF-M 8005PF-L 7705PFT-S 9500PFXS-06 6005PFXS 9905PFL 7705PFT-M 9500PFM 8005PF-XL 6005PF-XL 7705PFTXS 9905PF-XS 8005PF-S 6005PF-S 9500PFM-08 9500PFXL 9905PFS B6005PFXL B6005PFXS 6005PFS 9905PF-S
CA32891-813CS 764.13 CAD
CA32891-813 CA47751-888 CA47751-887 47746-090 47748-300 47746-092 47746-094 32891-815 CA47742-970 CA47742-972 CA47742-974 CA47746-946 CA47742-976 CA47742-968 47746-940 CA32891-833 47748-298 CA32891-831 47748-294 47751-892 CA32891-837 47751-890 CA32891-835 47746-942 47746-944 47746-946 47747-232 47746-948 47742-968 CA47746-086 47742-976 CA47746-088 47742-974 47746-950 47742-972 47746-952 47742-970 47748-306 CA32891-808 CA32891-809 CA47751-892 47748-308 CA47751-890 47748-304 28888-306 47746-954 47746-956 47751-887 47751-888 47746-088 CA32891-811 CA47746-090 CA47746-092 CA47746-094
N-DEX™ Ambidextrous Powder-Free Nitrile Gloves, Showa
Gloves
Gloves are constructed of puncture-resistant 100% nitrile which resists a broad range of chemicals and provides protection from intermittent contact and splashes.
  • Ambidextrous
  • Powder-free
  • Rolled cuffs
  • Smooth finish
  • Green gloves offer high visibility
Industrial-grade gloves are suitable for automotive, food, industrial, pharmaceutical, and laboratory applications. Gloves offer tactile sensitivity, comfort, flexibility, and dexterity. N-DEX™ Free and N-DEX™ Free Ultimate are accelerator-free.
Ordering information: All styles feature global sizing; for assistance in choosing the correct glove size, contact your Avantor sales representative.