Dichloromethane ≥99.5% stabilized ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Warning
|
Stabilization:
Stabilised with cyclohexene
LP300-19
LP300-4
CABJLP300D19EA
1243.29
CAD
CABJLP300D19
CABJLP300-4
Dichloromethane ≥99.5% stabilized ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Dichloromethane
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Formula:
CH₂Cl₂ Boiling Pt: 40 °C Melting Pt: –95 °C Density: 1.33 g/cm³ Flash Pt: Does not flash Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III Merck Index: 12,06140 |
|
Specification Test Results
Appearance | Passes test |
Assay by GC Analysis | 99.5 % min. |
Color (APHA) | 10 max. |
Free Halogens | Passes test |
Residue after Evaporation | 0.002 % max. |
Titrable Acid | 0.3 µeq/g max. |
Water by Karl Fischer Titration | 0.02 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...