Dichloromethane, anhydrous 99,9 Stabilised, BioSyn® Biochemistry, water_low_applications, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Warning
|
Synonyms:
Methylene chloride
Stabilization:
stab
BB301-4
BC301-200
CABJBB301-4CS
950.83
CAD
CABJBB301-4
BJBC301-200
BJCS301-56
Dichloromethane, anhydrous 99,9 Stabilised, BioSyn® Biochemistry, water_low_applications, Burdick & Jackson™
Dichloromethane
BioSyn products are ideal for DNA, RNA and oligonucleotide synthesis. All solvents and regents are filtered (0.2-micron) and tested for oxidation reage.
- Ultra-low water specifications
- Low amine specifications
- Low particulate levels
- High solvent purity
Formula:
CH₂Cl₂ MW: 84.93 g/mol Boiling Pt: 39.8 Melting Pt: -95 Density: 1.33 g/cm³ Flash Pt: Does not flash Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1 III Merck Index: 12,06140 |
|
Specification Test Results
Acidity (as HCl) | 1 mg/L max. |
Chloride | 10 mg/L max. |
GC Analysis | 99.9 % min. |
Refractive Index (20ºC) | 1.4236 - 1.4246 |
Residue | 1 mg/L max. |
UV Absorbance @ 240 nm | 0.1 AU max. |
UV Absorbance @ 250 nm | 0.01 AU max. |
UV Cutoff | 233 nm max. |
Water by Karl Fischer Titration | 20 ppm max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...