N,N-Dimethylformamide 99,9, BioSyn® Biochemistry, water_low_applications, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF
BB075-4
BN075-20
BN075-20
BC075-200
CABJBB075-4CS
3013.98
CAD
CABJBB075-4
CABJBN075-20
BJBN075-20
BJBC075-200
N,N-Dimethylformamide 99,9, BioSyn® Biochemistry, water_low_applications, Burdick & Jackson™
N,N-Dimethylformamide
BioSyn« products are ideal for DNA, RNA and oligonucleotide synthesis. All solvents and regents are filtered (0.2-micron) and tested for oxidation reage.
- Ultra-low water specifications
- Low amine specifications
- Low particulate levels
- High solvent purity
Formula:
HCON(CH₃)₂ MW: 73.09 g/mol Boiling Pt: 153 Melting Pt: -61 Density: 0.949 g/cm³ (20 °C) Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3 III REACH: 01-2119475605-32 Merck Index: 13,03269 |
|
Specification Test Results
Amines (as dimethylamine) | 3 ppm max. |
GC Analysis | 99.9 % min. |
Residue | 5 mg/L max. |
UV Absorbance @ 275 nm | 0.3 AU max. |
UV Absorbance @ 300 nm | 0.05 AU max. |
UV Absorbance @ 350 nm | 0.005 AU max. |
UV Absorbance @ 400 nm | 0.005 AU max. |
UV Cutoff | 268 nm max. |
Water by Karl Fischer Titration | 150 ppm max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...