Dichloromethane ≥99.5% stabilized ACS, VWR Chemicals BDH®

Supplier: VWR International
Warning

BDH1113-19L BDH1113-204L BDH1113-4LG
BDH1113-19LEA 1172.42 CAD
BDH1113-19L BDH1113-204L BDH1113-4LG
Dichloromethane ≥99.5% stabilized ACS, VWR Chemicals BDH®
Dichloromethane

Clear liquid. Contains cyclohexene as a preservative.

Formula: CH₂Cl₂
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear
Water (K.F.) Less than 0.020%
Residue Less than 0.002% (0.0010 g/76 mL)
GC Purity Greater than 99.5% (Typical - 99.9+%)
Color (APHA) Less than 10
Titrable acid Less than 0.3 eq/g (No more than 0.90 mL of 0.01N sodium hydroxide)
Free halogens Passes test (Lower layer should not show a violet tint)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR