N-Ethyldiisopropylamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA, Ethyldiisopropylamine
A11801-AE
A11801-AP
CAAAA11801-0FEA
812.28
CAD
CAAAA11801-0F
CAAAA11801-AE
CAAAA11801-AP
N-Ethyldiisopropylamine 99%
N-Ethyldiisopropylamine
Liquid. Flammable. Corrosive.
Purity: 99%
CAS No.: 7087-68-5
Molecular Formula: C8H19N
Formula Weight: 129.25
Flash Point: 8°(46°F)
Physical Form: Liquid
Density: 0.747
Refractive Index: 1.4135
Appearance: Colourless to pale yellow
Melting Point: -46°
Boiling Point: 126-127°
Beilstein Reference: 605301
UN No.: 3488
MDL No.: MFCD00008868
Formula:
C₈H₁₉N Boiling Pt: 126…127 °C Melting Pt: –46 °C Density: 0.747 Flash Pt: 8 °C(46 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008868 CAS Number: 7087-68-5 EINECS: 230-392-0 UN: 3384 ADR: "6.1",I |
Specification Test Results
Assay (by Metals Basis) | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...