Dysprosium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

Synonyms: Didysprosium trioxide, Dysprosium oxide

11319-09 11319-18 11319-30
CAAA11319-09EA 101.75 CAD
CAAA11319-09 CAAA11319-18 CAAA11319-30
Dysprosium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Dysprosium (III) oxide

Powder. Hygroscopic. Purity based on rare earth contaminates only.


In cermets used as nuclear reaction control rods


In cermets used as nuclear reaction control rods

Formula: Dy₂O₃
MW: 373 g/mol
Melting Pt: 2340 °C
Density: 7.81
Storage Temperature: Ambient
MDL Number: MFCD00010986
CAS Number: 1308-87-8
EINECS: 215-164-0
Merck Index: 14,3482

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR