3-Chloro-2,4,5,6-tetrafluoropyridine ≥98.0%

Supplier: TCI America
Warning

C1549-5G C1549-25G
TCC1549-005GEA 67.94 CAD
TCC1549-005G TCC1549-25G
3-Chloro-2,4,5,6-tetrafluoropyridine ≥98.0%
3-Chloro-2,4,5,6-tetrafluoropyridine

CAS Number: 1735-84-8
MDL Number: MFCD00010624
Molecular Formula: C5ClF4N
Molecular Weight: 185.51
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 71
Flash Point (°C): 113
Specific Gravity (20/20): 1.64

Formula: C₅ClF₄N
Boiling Pt: 71 °C
Density: 1.64 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00010624
CAS Number: 1735-84-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=851df90a532305337d9ffdf0d3bde3e4ed2b5958&vid=851df90a532305337d9ffdf0d3bde3e4ed2b5958&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR