1,3,5-Cyclohexanetriol (cis and trans mixture) ≥95.0%
Supplier: TCI America
C1051-25G
C1051-5G
TCC1051-025GEA
454.57
CAD
TCC1051-025G
76219-662
1,3,5-Cyclohexanetriol (cis and trans mixture) ≥95.0%
1,3,5-Cyclohexanetriol (cis and trans mixture)
CAS Number: 2041-15-8
MDL Number: MFCD00070580
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol |
MDL Number:
MFCD00070580 CAS Number: 2041-15-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...