CELLCOAT® Protein Coated Flasks, Greiner Bio-One
Supplier: Greiner Bio-One
658940 658950 661940 661950 690940 690950 690950-010 658950-005 690940-010 658940-005
82050-862CS 1362.54 CAD
82050-862 82050-864 82050-884 82050-886 82051-084 82051-086 75795-994 75795-996 75796-004 75796-006
CELLCOAT® Protein Coated Flasks, Greiner Bio-One
Flasks Cell Culture Flasks
These convenient, ready-to-use tissue culture flasks are coated with extra-cellular matrix proteins (ECM) or synthetic peptides.
  • High-profile design
  • Nonpyrogenic
  • Free of RNase and DNase
  • Convenient and ready to use
Coating provides an inert scaffolding that helps regulate cell growth and proliferation. It simulates the cells' natural environment, which facilitates adherence, differentiation, proliferation, and longevity in many cell types. Flasks also feature a canted neck, plug seal, "Best Before" date, and a lot number on each pack. Flasks are nonpyrogenic and DNase- and RNase-free. All 650 mL flasks feature a high profile design.