VWR® CERTICLEAN® Cleanroom Nitrile Gloves, Class 10
Supplier: VWR International
CA40101-352 CA40101-353 CA40101-354 CA40101-356 CA40101-358 CA40101-357
CA40101-352CS 713.36 CAD
CA40101-352 CA40101-353 CA40101-354 CA40101-356 CA40101-358 CA40101-357
VWR® CERTICLEAN® Cleanroom Nitrile Gloves, Class 10
Gloves
Ambidextrous, powder-free gloves with beaded cuffs for strength and reduction of cuff roll down.
  • Superior strength and puncture resistance
  • Certificates of analysis available
  • Quality data available
Nitrile is superior in strength and puncture resistance compared to other glove compounds. Film thickness designed for comfort and tactile sensitivity. Gloves are 6.3 mil thick at fingers and 5.1 mil thick at palms. Textured fingertips for a reliable grip. Uncompromising manufacturing processes yield consistently low particle counts. Proprietary nitrile, silicone-free formulation, and specialized manufacturing processes result in consistently low levels of ionic extractables and nonvolatile residue (NVR) transfer. In-use static dissipative properties superior to natural rubber latex.
The 30.5 cm (12") length and tapered cuff provide more coverage over the garment sleeve.
Packaging: Packed in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Packaged in cleanroom-compatible polyethylene, 100 gloves per double poly bag, 10 bags per case.