TouchNTuff® 92-500 Nitrile Gloves, Powdered, Ansell
Supplier: Ansell Canada
9250011070 9250011080 9250011090 9250011100
CA32490-000CS 353.79 CAD
CA32490-000 CA32490-001 CA32490-002 CA32490-003
TouchNTuff® 92-500 Nitrile Gloves, Powdered, Ansell
Gloves
Made of 100% nitrile, TouchNTuff® 92-500 disposable gloves are thin yet strong, providing both tactile sensitivity and splash resistance against hazardous chemicals.
  • Excellent fingertip sensitivity
  • Exceptional durability
  • Lightly powdered and lightweight
  • Silicone-free
They offer three times the puncture resistance of standard latex or vinyl gloves. These gloves combine excellent fingertip sensitivity with exceptional durability to give a high level of protection against grease and oil. They are lightly powdered, lightweight and comfortable to wear.