Nitrile Gloves, Kimtech™ G3 NXT™, Kimberly-Clark
Supplier: Microflex
62990 62991 62992 62995 62994 62992
CA14214-536CS 591.54 CAD
CA14214-536 CA14214-538 CA14214-540 CA14214-543 CA14214-544 14214-540
Nitrile Gloves, Kimtech™ G3 NXT™, Kimberly-Clark
Gloves
These powder-free nitrile gloves feature textured fingertips to facilitate control and grip.
  • Powder free
  • Textured fingertips for control and grip
  • Ambidextrous
  • Beaded cuff
AQL 1.5.
Certifications: Recommended for ISO Class 4 (FED-STD-209E Class 1/M1.5) or higher cleanroom environments.
Ordering information: Packaged in a Class 100 cleanroom. Packed 100 gloves per double poly bag with case liner.