CE5 System Nitrile Gloves, Microflex®, Ansell
Supplier: Microflex
CE5-755-M CE5-755-XL CE5-755-XS
CA32916-762CS 500.26 CAD
CA32916-762 CA32916-764 CA32916-766 CA32916-768 75816-068
CE5 System Nitrile Gloves, Microflex®, Ansell
Gloves
These ambidextrous gloves are for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) controlled environments.
  • Silicone-free
  • Powder-free
  • Superb tactile sense and comfort
  • Low surface particulates
These gloves produce only very low levels of surface particulates, ionic extractables, and ono-volatile residues.
Certifications: Lot-specific Certificate of Analysis available upon request.
Packaging: Packed 100 gloves per vacuum-sealed double poly bag, 10 poly bags per case.
Caution: Components used in making these gloves may cause allergic reactionsMore Product Information