4-Cyanopyridine-N-oxide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
C0765-25G
C0765-500G
TCC0765-025GEA
199.08
CAD
TCC0765-025G
TCC0765-500G
4-Cyanopyridine-N-oxide ≥98.0% (by titrimetric analysis)
4-Cyanopyridine N-oxide
CAS Number: 14906-59-3
MDL Number: MFCD00006205
Molecular Formula: C6H4N2O
Molecular Weight: 120.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 230
Other Product Names / Synonyms: Isonicotinonitrile N-Oxide
Formula:
C₆H₄N₂O Melting Pt: 230 °C |
MDL Number:
MFCD00006205 CAS Number: 14906-59-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...