Color Standard, APHA / HzAen (Platinum-Cobalt), Color = 500, Ricca Chemical Company
Supplier: Ricca Chemical
R2230000-1C
R2230000-500C
RC223032EA
1641.62
CAD
RC223032
RC223016
Color Standard, APHA / HzAen (Platinum-Cobalt), Color = 500, Ricca Chemical Company
Standards
Colour Reference and Spectroscopy Standards
APHA and ASTM
Stock platinum-cobalt formula, 500mg/L platinum.