Chromicsulfuric acid solution for cleaning of laboratory glassware
Supplier: Ricca Chemical
|
Danger
|
R2150000-1C1
R2150000-2.5D
RC215032EA
192.76
CAD
RC215032
RC21505PT
Chromicsulfuric acid solution for cleaning of laboratory glassware
Chromicsulfuric acid solution
For cleaning glassware.
Boiling Pt:
~ 300 °C Density: 1.8000 g/cm³ Storage Temperature: Ambient |
UN:
1830 ADR: 8,II |
Specification Test Results
Appearance | Dark brown, viscous liquid, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...