Chloride Standard, 1000 ppm Cl⁻ (0.0282 Normal), Ricca Chemical Company

Supplier: Ricca Chemical

R1955000-120A R1955000-500A R1955000-250A R1955000-1A R1955000-4A
RC1955-4EA 90.06 CAD
RC1955-4 RC1955-16 RC1955-8 RCSC023100-5A RCR1955000-1A RCR1955000-4A
Chloride Standard, 1000 ppm Cl⁻ (0.0282 Normal), Ricca Chemical Company
Standards Anionic Standards Chloride Standards

Chloride Standard, 1000 ppm Cl⁻ (0.0282 Normal) for use with ISEs, ion chromatography, and other anion analysis.



Specifications

Appearance: Colorless Liquid

Assay: 999-1001 ppm Cl


Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR