Calmagite 0.1% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical

R1830000-500A R1830000-120A
RC183016EA 112.18 CAD
RC183016 RC1830-4
Calmagite 0.1% (w/v) in aqueous solution indicator
Calmagite

APHA, for water hardness.

Formula: C₁₇H₁₄N₂O₅S
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011656
CAS Number: 3147-14-6

Order Now

Specification Test Results

Appearance Dark red, characteristic Calmagite odor
Suitability for Use Wine Red (Excess Ca/Mg) - Pure Blue (Excess EDTA)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR