Duro Task™ Supported Natural Rubber Gloves, Honeywell Safety
Supplier: Honeywell Safety
Duro Task™
NF14/7S NF14/9L NF14/9L NF14/8M NF14/7S NF14/10XL NF14/11XXL NF14/8M NF14/10XL
CA89022-118CS 1317.37 CAD
CA89022-118 89022-118 CA89022-116 CA89022-114 89022-116 89022-114 CA89022-120 89022-122 89022-120
Duro Task™ Supported Natural Rubber Gloves, Honeywell Safety
Gloves
Economical gloves with rubber coating for general maintenance, manufacturing, and assembly applications.
  • Seamless liner provides added comfort and lessens hand fatigue
  • Cuff trim size is color-coded for easy identification
Textured finish provides sure grip on objects. Fingertip and thumb crotch coating helps minimize wear on high-stress areas.