VWR® Cleanroom Binders
Supplier: VWR
VWR®
P1114-3 P1114-1 BIN85111P P1114-2
51280-752CS 1963.99 CAD
51280-752 51280-796 51280-826 51280-756
VWR® Cleanroom Binders
Binders and Indexes
These three-ring binders are constructed of antistatic 75-gauge, high-density polyethylene.
  • Polyethylene fabrication
  • Transparent overlay on front and spine allows for identification sheet insertion
  • Multiple ring sizes available
Choose from strong metal or plastic rings.
Tab dividers 51280-832 are designed for cleanroom binders. Dividers are three-hole punched with a 1/5 cut tab.
For 21.6Wx27.9Lcm (8.5x11") paper.
Ordering information: Four-ring binders, custom sizes, and custom colors are also available; contact your Avantor sales representative for more information.