TouchNTuff® 69-318 Natural Rubber Latex Disposable Gloves, Ansell
Supplier: Ansell Healthcare
6931800S 693180XL 6931800L
36888-210CS 386.8 CAD
36888-210 36888-216 36888-212 36888-214
TouchNTuff® 69-318 Natural Rubber Latex Disposable Gloves, Ansell
Gloves
SType 69-318 gloves are 100% natural rubber latex.
  • Powder-free
  • Ambidextrous
  • Textured grip
Gloves are suitable for laboratory analysis/technical work, food processing and handling, quality control, electronics assembly, as well as light-duty maintenance and cleanup.
Certifications: Gloves qualify as Class I Medical Devices. Component materials comply with FDA regulations for food contact.
Caution: This product contains natural rubber latex which may cause allergic reactions in some individuals.