Cyclohexane ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: HexahydrobenzeneHexamethylene

Management of Change (MOC) category = NI

9258-03 9258-03 BAKR9258-H1 9258-H1
CAJT9258-3CS 3261.75 CAD
CAJT9258-3 JT9258-H1 CAJT9258-H1 JT9258-3
Cyclohexane ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Cyclohexane

For organic residue analysis.

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C (closed cup)
MDL Number: MFCD00003814
CAS Number: 110-82-7
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://ca.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤10
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (C₆H₁₂) (by GC, corrected for water) ≥99.0 %
Color (APHA) ≤10
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR