Chloroform ≥99.8% (by GC, corrected for water content) stabilized, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: VWR International
Danger


Stabilization: Stabilised with ethanol 1 %

CAJT9180-8 BAKR9180-R BAKR9180-01 BAKR9180-22 BAKR9180-03 BAKR9180-33 BAKR9180-05
CAJT9180-8EA 7391.95 CAD
CAJT9180-8 CAJT9180-R CAJT9180-1 CAJT9180-22 CAJT9180-3 CAJT9180-33 CAJT9180-5 CAJT9180-54
Chloroform ≥99.8% (by GC, corrected for water content) stabilized, BAKER ANALYZED® ACS, J.T.Baker®
Chloroform

Stabilized with ~1% ethanol. Lot analysis on label.

Formula: CHCl₃
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water) ≥99.8 %
Acetone, Aldehydes (as (CH₃)₂CO) Passes Test
Acid and Chloride Passes Test
Color (APHA) ≤10
Ethanol (CH₃CH₂OH)(by GC) 0.7 - 1.0 %
Free Chlorine Passes Test
Lead (Pb) ≤0.050 ppm
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Suitability for Use in Dithizone Tests Passes Test
Water (by KF, coulometric) ≤0.01 %
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR