Chlorobenzene ≥99.5% (by GC), BAKER ANALYZED® ACS

Supplier: VWR International
Warning

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

Management of Change (MOC) category = N

CAJT9179-8 BAKR9179-03 9179-01
CAJT9179-8EA 2982.95 CAD
CAJT9179-8 CAJT9179-1 CAJT9179-3 JT9179-1
Chlorobenzene ≥99.5% (by GC), BAKER ANALYZED® ACS
Chlorobenzene

Lot analysis on label.

Formula: C₆H₅Cl
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.11 g/cm³ (20 °C)
Flash Pt: 28 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₆H₅Cl) (by GC) ≥99.5 %
ACS - Acidity (as HCl) ≤0.015 %
ACS - Color (APHA) ≤30
ACS - Residue after Evaporation ≤0.010 %
ACS - Titrable Acid (meq/g) ≤0.004


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR