Carbon disulfide 99,9 GC, corrected for water content, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


M.O.C. Category N

BAKR9172-22 BAKR9172-05
CAJT9172-22CS 2477.44 CAD
CAJT9172-22 CAJT9172-5
Carbon disulfide 99,9 GC, corrected for water content, BAKER ANALYZED® ACS
Carbon disulphide

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46.2
Melting Pt: -111.6
Density: 1.2632 g/cm³ (20 °C)
Flash Pt: –30 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3 I
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CS₂) (by GC, corrected for water) ≥99.9 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Hydrogen Sulfide (H₂S) Passes Test
Sulfur Dioxide (SO₂) Passes Test
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR