Copper(II) chloride dihydrate 99.0-101.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

Management of Change (MOC) category = N

BAKR1792-01 BAKR1792-04 BAKR1792-05
CAJT1792-1CS 2804.37 CAD
CAJT1792-1 CAJT1792-4 CAJT1792-5
Copper(II) chloride dihydrate 99.0-101.0%, BAKER ANALYZED® ACS, J.T.Baker®
Copper (II) chloride dihydrate

Crystals. Lot analysis on label.

Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 °C
Density: 2.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CuCl₂·2H₂O) 99.0 - 101.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - pH of 5% Solution at 25°C 2.0 - 4.0
ACS - Nitrate (NO₃) ≤0.015 %
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Iron (Fe) ≤0.005 %
ACS - Nickel (Ni) ≤0.01 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR