Latex Gloves, Ambidextrous, Kimtech™ G3, Kimberly-Clark
Supplier: Microflex
56813 56814 56816
CA32933-871CS 522.17 CAD
CA32933-871 CA32933-873 CA32933-875 CA32933-877
Latex Gloves, Ambidextrous, Kimtech™ G3, Kimberly-Clark
Gloves
Designed for critical cleanroom environments, these specially processed gloves are washed multiple times in ultrapure deionized water to ensure consistent control of particles and extractables.
  • Recommended for ISO Class 4 (FED-STD-209E Class 1/M1.5) or higher cleanrooms
  • Textured palm and fingers
  • Ambidextrous
  • AQL 1.5
Gloves offer exceptional wet/dry grip without compromising tactile sensitivity. The tapered, beaded cuff makes donning easier, reduces rolldown, enhances cuff strength, and forms a better seal on the garment.
Caution: This productMore Product Information