Calcium pantothenate 90.0-100.0% USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Calcium D-Pantothenate, D-Pantothenic acid calcium salt, D-Calcium pantothenate, Calcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate, Vitamin B5 calcium salt, Calcium D-(+)-pantothenate

Management of Change (MOC) category = R

BAKR1443-03 BAKR1443-08
CAJT1443-3EA 259.42 CAD
CAJT1443-3 CAJT1443-8
Calcium pantothenate 90.0-100.0% USP, J.T.Baker®
Calcium pantothenate
Formula: C₁₈H₃₂CaN₂O₁₀
MW: 476.54 g/mol
Melting Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00002766
CAS Number: 137-08-6
EINECS: 205-278-9

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Identification A Passes Test
Identification B Passes Test
Identification C (Specific Rotation [Å]²⁵ ^D (+)) 25.0 - 27.5 Degree
Alkalinity Passes Test
Loss on Drying ≤5.0 %
Assay (C₁₈H₃₂CaN₂O₁₀)(dried basis) 98.0 - 102.0 %
Content of Calcium (Ca)(dried basis) 8.2 - 8.6 %
Solution Test Passes Test
Only Class 2 solvents (Methanol, Dichloromethane) are likely to be
present. All are below Option 1 limits.
Meets Reagent Specifications for testing USP/NF monographs
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR