Latex Gloves, Hand-Specific, Kimtech™ G3, Kimberly-Clark
Supplier: Microflex
56827 56828 56830 56831 56832 56833 56825 56834
CA32933-716CS 668.86 CAD
CA32933-716 CA32933-718 CA32933-720 CA32933-724 CA32933-726 CA32933-728 CA32933-730 CA32933-734 CA32933-736 CA32933-738 CA32933-740 CA32933-742 CA32933-811 CA32933-813 32933-722
Latex Gloves, Hand-Specific, Kimtech™ G3, Kimberly-Clark
Gloves
Designed for critical cleanroom environments.
  • Double-bagged to ensure cleanliness
  • Textured palms and fingertips
Gloves offer exceptional wet/dry grip without compromising tactile sensitivity. The tapered, beaded cuff reduces rolldown to make donning easier, enhances cuff strength, and forms a better seal on the garment. AQL 1.5.
Specially processed gloves are washed multiple times in ultrapure deionized water to ensure consistent control of particles and extractables.
Certifications: Recommended for ISO Class 4 or higher cleanroom environments.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established. This product should not be worn by or exposed to individuals allergic to natural rubber latex.