Calcium chloride, anhydrous 96,0 Argentometric titration, BAKER ANANYZED™ ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

M.O.C. Category N

BAKR1311-07 BAKR1311-05 1311-01
CAJT1311-7EA 3516.59 CAD
CAJT1311-7 CAJT1311-1 CAJT1311-5 JT1311-1
Calcium chloride, anhydrous 96,0 Argentometric titration, BAKER ANANYZED™ ACS
Calcium chloride

Desiccant, 20 mesh. Lot analysis on label.

Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CaCl₂) (by Ag titrn) ≥96.0 %
Titrable Base (meq/g) ≤0.006
Mesh - On U.S. No. 20 Sieve ≤10 %
Mesh - Thru U.S. No. 200 Sieve ≤30 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR