Citric acid, anhydrous 99.5-100.5% (by anhydrous basis), powder USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = R

BAKR0122-07 BAKR0122-05 0122-01 BAKR0122-R
CAJT0122-7EA 3585.42 CAD
CAJT0122-7 CAJT0122-1 CAJT0122-5 JT0122-1 CAJT0122-R
Citric acid, anhydrous 99.5-100.5% (by anhydrous basis), powder USP, J.T.Baker®
Citric acid

Avantor excipients are used during the formulation process in a wide range of applications. Avantor provides high-purity and performance excipients that serve as fillers, binders, disintegrants, lubicants, flavors, emulsifiers and preservatives. All the products are cGMP produced and meet USP grade standards.

Formula: HOC(COOH)(CH₂COOH)₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 149…151 °C
Density: 1.44 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis) 99.5 - 100.5 %
USP - Clarity of Solution Passes Test
USP - Color of Solution Passes Test
USP - Identification Passes Test
USP - Residue on Ignition ≤0.1 %
USP - Oxalic Acid ≤0.036 %
USP - Sulfate (SO₄) ≤0.015 %
USP - Readily Carbonizable Substances Passes Test
USP - Identification C: Water (H₂O) ≤1.0 %
Must be subjected to further processing during the preparation of
injectable dosage forms
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Meets Reagent Specifications for testing USP/NF monographs
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR