Ultra One® Latex Gloves, Microflex®, Ansell
Supplier: Microflex
UL-315-S UL-315-M UL-315-L UL-315-XL UL-315-XL UL-315-S UL-315-M UL-315-L
CA32916-516CS 379.2 CAD
CA32916-516 CA32916-518 CA32916-520 CA32916-522 32916-522 32916-516 32916-518 32916-520
Ultra One® Latex Gloves, Microflex®, Ansell
Gloves
These gloves have extended, beaded cuffs for increased protection over the wrist and forearm.
  • A standard for high-risk applications
  • Fingertip texture allows for better grip in wet and dry conditions
Packaging: Packaged 50 gloves per dispenser box.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex sensitized individuals has not been established.