Stainless Steel Dressing Jars, Polar Ware
Supplier: Polar Ware
3J 4J 1J 2J
36312-006EA 543.65 CAD
36312-006 36312-007 36312-004 36312-005
Stainless Steel Dressing Jars, Polar Ware
Jars General Use Jars
Stainless steel dressing jars clean easily and sterilize quickly
Stain-proof and long lasting. Will not chip, break or corrode.