KIMAX® Dropping Bottles, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
15035-125 15035-15 15035-30 15035-60
89001-560CS 1731.68 CAD
89001-560 89001-562 89001-564 89001-566
KIMAX® Dropping Bottles, Kimble®, DWK Life Sciences
Bottles Dropping Bottles
These borosilicate glass bottles have 1 ml dropper capacity and include a medium length [ST] pipet dropper with a rubber nipple.
The 30 and 60 ml sizes are KG-33 ASTM 438, Type 1, Class A glass and meet Federal Specification NNN-B-1194, Type II, Class 2, Grade B requirements. The 125 ml size is KG-35 ASTM, 438 Type 1, Class B glass.
Bottles are suitable for use in determining the oil absorption of pigments (ASTM D281).