VWR® CERTICLEAN® Cleanroom Latex Gloves, Ambidextrous, Class 10
Supplier: VWR International
CA40101-146 CA40101-148 CA40101-152 CA40101-150
CA40101-146CS 731.33 CAD
CA40101-146 CA40101-148 CA40101-152 CA40101-150
VWR® CERTICLEAN® Cleanroom Latex Gloves, Ambidextrous, Class 10
Gloves
Ambidextrous, powder-free, natural-colored gloves with beaded cuffs for strength and reduction of cuff roll down.
  • Powder-free, natural rubber latex
  • Tapered, beaded cuff
  • Certificates of analysis available
  • Quality data available
Uncompromising manufacturing processes consistently yield exceptionally low particle counts. Specialized manufacturing processes and silicone-free material minimize ionic contaminants and nonvolatile residue (NVR) transfer.
The length and tapered cuff provide more coverage over the garment sleeve. Film thickness designed forMore Product Information