VWR® PE Stoppers, Flathead
Supplier: Avantor
VWR®
011.432.02 011.432.08 011.432.06 011.432.04 011.432.05 011.432.07 011.432.03
77441-900CS 279.78 CAD
77441-900 77441-902 77441-896 77441-898 77441-892 77441-894 77441-890
VWR® PE Stoppers, Flathead
Stoppers PE/LDPE/HDPE Stoppers
Solid PE stoppers with flat head.
  • Rings along the plug provide an effective seal
  • Wide flange offers protection in the event of accidental tipping
For use with volumetric flasks and measuring cylinders.