VWR® PTFE Stoppers, [ST]
Supplier: Avantor
VWR®
011.278.08 011.278.00 011.278.07 011.278.04
77441-888CS 3391.96 CAD
77441-888 77441-884 77441-886 77441-882
VWR® PTFE Stoppers, [ST]
Stoppers PE/LDPE/HDPE Stoppers
Solid PTFE stoppers with T-shaped head.
  • T-handles for easy on and off
For use with round bottom flasks, Erlenmeyer flasks, and separating funnels.