Cyclohexanone ≥99% (by GC), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Pimelic Ketone

CX2335-20 CX2335-5 CX2335-1 CX2335-7 CX2335-1
CACX2335-20EA 11667 CAD
CACX2335-20 CACX2335-5 CACX2335-1 CACX2335-7 CATXCX2335-1
Cyclohexanone ≥99% (by GC), GR ACS, Supelco®
Cyclohexanone

Meets ACS Specifications

Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –31 °C
Density: 0.95 g/cm3 (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Expiration date Exp 24 months from receipt date
Assay (GC) 99.0% min
Color (APHA) at time of manufacture 10 max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 0.05% max
Water (H2O) 0.05% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR