Chloroform ≥99.8% stabilised for HPLC, Supelco®

Supplier: MilliporeSigma
Danger


Stabilization: Stabilised with ethanol

CX1058-6 CX1058-1 CX1058P-1 CX1058-1
CACX1058-6EA 227 CAD
CACX1058-6 CACX1058-1 CACX1058P-1 EM-CX1058-1
Chloroform ≥99.8% stabilised for HPLC, Supelco®
Chloroform

HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: CHCl₃
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (CHCl3 + C2H5OH) 99.8% min
Acetone and Aldehydes (about 0.005%) To pass test
Acid and chloride To pass test
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Density at 25°C 1.471-1.476g/mL
Ethanol 0.7-1.0% (v/v)
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 1.0ppb max
Free chlorine (Cl) To pass test
Infrared spectrum Conforms to standard
Lead (Pb) 0.05ppm max
Refractive index (n 20/D) 1.442-1.446
Residue after evaporation 2ppm max
Substances darkened by sulfuric acid To pass test
Suitability for dithizone tests To pass test
UV Abs. at 245 nm 1.00AU max
UV Abs. at 255 nm 0.25AU max
UV Abs. at 260 nm 0.15AU max
UV Abs. at 270 nm 0.05AU max
UV Abs. at 290 - 400 nm 0.01AU max
UV Cut-off 244nm max
Water (H2O) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR