Chloroform ≥99.9% stabilised, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Stabiliser

CX1054-NP20 CX1054-1 CX1054-6 CX1054P-1
CACX1054-NP20EA 1814 CAD
CACX1054-NP20 CACX1054-1 CACX1054-6 CACX1054P-1
Chloroform ≥99.9% stabilised, OmniSolv®, Supelco®
Chloroform

For spectrophotometry, HPLC, GC, and residue analysis. Stabilized with amylene. UV cutoff 245nm. Certificate of Analysis on label.

Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: -63
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Capillary ECD responsive substances (as PCNB) 2ng/L max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 250ppt max
Form Clear liquid
Free chlorine (Cl) To pass test
Identity (IR-spectrum) Conforms
Residue after evaporation 1ppm max
Titrable acid 0.2µeq/g max
UV Abs. at 245 nm 1.00AU max
UV Abs. at 250 nm 0.30AU max
UV Abs. at 260 nm 0.04AU max
UV Abs. at 270 nm 0.005AU max
UV Abs. at 300 nm 0.005AU max
UV Cut-off 245nm max
Water (H2O) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR