Chloroform ≥99.8% stabilised for HPLC, Supelco®

Supplier: MilliporeSigma
Danger

CX1050-1 CX1050P-1
CACX1050-1CS 1806 CAD
CACX1050-1 CACX1050P-1
Chloroform ≥99.8% stabilised for HPLC, Supelco®
Chloroform

HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: CHCl₃
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.8% min
Acetone and Aldehydes (about 0.005%) To pass test
Acid and chloride To pass test
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Free chlorine (Cl) To pass test
Infrared spectrum Conforms to standard
Lead (Pb) 0.05ppm max
Residue after evaporation 5ppm max
Suitability for dithizone tests To pass test
UV Abs. at 245 nm 1.00AU max
UV Abs. at 255 nm 0.25AU max
UV Abs. at 260 nm 0.15AU max
UV Abs. at 270 nm 0.05AU max
UV Abs. at 290 - 400 nm 0.01AU max
UV Cut-off 245nm max
Water (H2O) 0.05% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR