1-Chlorobutane ≥99.5%, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: n-Butyl Chloride

CX0914-1 CX0914-6
CACX0914-1CS 3233 CAD
CACX0914-1 CACX0914-6
1-Chlorobutane ≥99.5%, OmniSolv®, Supelco®
1-Chlorobutane

For spectrophotometry and chromatography. Filtered through a 0.2µm element. UV cutoff 220nm. Lot analysis on label.

Formula: CH₃(CH₂)₃Cl
MW: 92.57 g/mol
Boiling Pt: 78.4 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.8862 g/cm³ (20 °C)
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3,II
Merck Index: 13,01559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.5% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 1.0ppb max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 1ppm max
UV Abs. at 220 nm 1.00AU max
UV Abs. at 225 nm 0.50AU max
UV Abs. at 233 nm 0.10AU max
UV Abs. at 240 nm 0.05AU max
UV Abs. at 270 nm 0.01AU max
UV Abs. at 300 nm 0.005AU max
UV Cut-off 220nm max
Water (H2O) 0.01% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR