Carbon disulfide ≥99.9%, OmniSolv® low in benzene, Supelco®

Supplier: MilliporeSigma

CX0397-6 CX0397-6
CACX0397-6CS 15741.32 CAD
CACX0397-6 EM-CX0397-6
Carbon disulfide ≥99.9%, OmniSolv® low in benzene, Supelco®
Carbon disulphide

Flammable.

Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46.2 °C (1013 hPa)
Melting Pt: –111.6 °C
Density: 1.2632 g/cm³ (20 °C)
Flash Pt: –30 °C
MDL Number: MFCD00011321
CAS Number: 75-15-0
UN: 1131
ADR: 3,I
Merck Index: 13,01821
https://ca.vwr.com/stibo/web/std.lang.all/53/94/785394.jpg

Order Now

Specification Test Results

Assay (GC) 99.9% min
Benzene 200ppb max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Other volatile impurities 0.5ppm max
Residue after evaporation 3ppm max
UV Abs. at 382 nm 1.00AU max
UV Abs. at 388 nm 0.50AU max
UV Abs. at 403 nm 0.10AU max
UV Abs. at 410 nm 0.05AU max
UV Abs. at 500 nm 0.01AU max
UV Cut-off 382nm max
Water (H2O) 0.03% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR